missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PNMA6A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560893
This item is not returnable.
View return policy
Description
Immunogen sequence: GPWNVVFVPR CSGEEFLGLG RVFHFPEQEG QMVESVAGAL GVGLRRVCWL RSIGQAVQPW VEAVRCQSLG VF Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 31%.
Specifications
| PNMA6A | |
| Polyclonal | |
| Unconjugated | |
| PNMA6A | |
| MA6; paraneoplastic antigen-like 6A related protein; paraneoplastic antigen-like protein 6A; paraneoplastic antigen-like protein 6C; paraneoplastic Ma antigen family member 6A; paraneoplastic Ma antigen family member 6C; PNMA6; PNMA6A; PNMA6C | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84968 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P0CW24 | |
| PNMA6A | |
| Recombinant protein corresponding to Human PNMA6A. Recombinant protein control fragment (Product #RP-104378). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction