Learn More
PNMA5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560796
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: QARSFSDSSP QTIQGGLPPL VKRRRLLGSE STRGEDHGQA TYPKAENQTP GREGPQAAGE ELGNEAGAGA MSHPK Highest antigen sequence identity to the following orthologs: Mouse - 39%, Rat - 45%.
This gene encodes a member of the paraneoplastic Ma antigen protein family. These proteins have been implicated in the development of paraneoplastic disorders resulting from an immune response directed against them. Paraneoplastic disorders are the result of an abnormal immune response to a tumor. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq, Oct 2011].Specifications
PNMA5 | |
Polyclonal | |
Unconjugated | |
PNMA5 | |
KIAA1934; Paraneoplastic antigen-like protein 5; paraneoplastic Ma antigen family member 5; PNMA5; Tumor antigen BJ-HCC-25 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114824 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96PV4 | |
PNMA5 | |
Recombinant protein corresponding to Human PNMA5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only