missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PNLIPRP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562530
This item is not returnable.
View return policy
Description
Immunogen sequence: KHMPGCEDLI TPLLKFNFNA YKKEMASFFD CNHARSYQFY AESILNPDAF IAYPCRSYTS Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 45%.
Specifications
| PNLIPRP3 | |
| Polyclonal | |
| Unconjugated | |
| PNLIPRP3 | |
| pancreatic lipase related protein 3; pancreatic lipase-related protein 3; PL-RP3; PNLIPRP3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 119548 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q17RR3 | |
| PNLIPRP3 | |
| Recombinant protein corresponding to Human PNLIPRP3. Recombinant protein control fragment (Product #RP-104266). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction