missing translation for 'onlineSavingsMsg'
Learn More

PNLDC1 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£149.00 - £364.00

Specifications

Antigen PNLDC1
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18681722
View Documents
Novus Biologicals
NBP2-94424-0.02ml
0.02 mL
£149.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18663652
View Documents
Novus Biologicals
NBP2-94424-0.1ml
0.1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

PNLDC1 Polyclonal antibody specifically detects PNLDC1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

PNLDC1
Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
dJ195P10.2, FLJ40240, MGC138451, poly(A)-specific ribonuclease (PARN)-like domain containing 1, poly(A)-specific ribonuclease PARN-like domain-containing protein 1
Recombinant fusion protein containing a sequence corresponding to amino acids 200-500 of human PNLDC1 (NP_775787.1). DEGVVVKKVSKQHRWYLQNTSCDRESCWKENILLSARGFSVFFQMLVKAQKPLVGHNMMMDLLHLHEKFFRPLPESYDQFKQNIHSLFPVLIDTKSVTKDIWKEMNFPRVSNLSEVYEVLNSDLNPTKNSGPEIVHASRCEKYVETKCPHEAAYDAFLCGSVLLKVAHLLLQKIYHIDPVPESSFPQYLDVLAPYVNQVNLIRAGVPKINFSGPDYPSIRPPILILSVKRWPGVSEQQVYHKFQNLCKFDVRRLTRSQFLLLTNKFKDARNILKEYRDHPTLCISLYRYWRHSPNVNCLLQ
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot 1:500-1:2000
Polyclonal
Purified
RUO
PBS (pH 7.3), 50% glycerol
154197
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.