Learn More
PNKD Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552695
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: GATANKASHN RTRALQSHSS PEGKEEPEPL SPELEYIPRK RGKNPMKA Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
Probable hydrolase that plays an aggravative role in the development of cardiac hypertrophy via activation of the NF-kappa-B signaling pathway.Specifications
PNKD | |
Polyclonal | |
Unconjugated | |
PNKD | |
2210013N15Rik; 2810403H05Rik; AI854243; brain protein 17; Brp17; DYT8; FKSG19; FPD1; hydrolase PNKD-like protein; KIAA1184; KIPP1184; LOC106780846; LOW QUALITY PROTEIN: probable hydrolase PNKD; MNCb-5687; MR1; MR-1; Myofibrillogenesis regulator 1; paroxysmal nonkinesigenic dyskinesia; paroxysmal nonkinesiogenic dyskinesia; paroxysmal nonkinesiogenic dyskinesia protein; PDC; PKND1; Pnkd; PNKD metallo-beta-lactamase domain containing; PNKD, MBL domain containing; probable hydrolase PNKD; probable hydrolase PNKD; LOW QUALITY PROTEIN: probable hydrolase PNKD; similar to brain protein 17 isoform 2; TAHCCP2; Trans-activated by hepatitis C virus core protein 2; UNQ2491/PRO5778 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25953 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N490 | |
PNKD | |
Recombinant protein corresponding to Human PNKD | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only