missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PNKD Polyclonal antibody specifically detects PNKD in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | PNKD |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS (pH 7.4) |
| Gene Accession No. | AAH36457.1 |
| Gene Alias | BRP17FKSG19, DKFZp564N1362, DYT8paroxysmal nonkinesiogenic dyskinesia, FPD1brain protein 17, KIAA1184Myofibrillogenesis regulator 1, KIPP1184probable hydrolase PNKD, MGC31943, MR1Paroxysmal nonkinesiogenic dyskinesia protein, MR-1Trans-activated by hepatitis C virus core protein 2, paroxysmal nonkinesigenic dyskinesia, PDCEC 3.-, TAHCCP2PKND1 |
| Host Species | Mouse |
| Immunogen | MR-1 (AAH36457.1, 1 a.a. - 385 a.a.) full-length human protein. MAAVVAATALKSRGARNARVLRGILAGATANKVSHNRTRALQSHSSSEGKEEPEPLSPELEYIPRKRGKNPMKAVGLAWYSLYTRTWLGYLFYRQQLRRARNRYPKGHSKTQPRLFNGVKVLPIPVLSDNYSYLIIDTQAQLAVAVDPSDPRAVQASIEKEGVTLVAILCTHKHWDHSGGNRDLSRRHRDCRVYGSPQDGIPYLTHPLCHQDVVSVGRLQIRALATPGHTQGHLVYLLDGEPYKGPSCLFSGDLLFLSGCGRTFEGNAETMLSSLDTVLGLGDDTLLWPGHEYAEENLGFAGVVEPENLARERKMQWVQRQRLERKGTCPSTLGEERSYNPFLRTHCLALQEALGPGPGPTGDDDYSRAQLLEELRRLKDMHKSK |
| Purification Method | Protein G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?