missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528853
This item is not returnable.
View return policy
Description
Recommended positive controls: A549, HeLa, HepG2. Predicted reactivity: Mouse (82%), Rat (81%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Anti-human Polynucleotide Kinase Mouse Monoclonal Antibody recognizes polynucleotide kinase (PNK), a 57 kDa nuclear enzyme implicated in DNA strand repair processes that occur during DNA replication and recombination, or in response to DNA damaging agents. PNK targets the DNA 5'-hydroxy and 3'-phosphate termini for phosphorylation and dephosphorylation events. During strand breakage in human cells, normal PNK associates with other nuclear proteins to prepare the DNA termini for the enzymatic actions of DNA polymerase-beta and DNA ligase III. PNK is a key functional component in maintaining the genetic stability of human cells.
Specifications
| PNK | |
| Polyclonal | |
| Unconjugated | |
| Pnkp | |
| 1810009G08Rik; 2'(3')-polynucleotidase; AOA4; bifunctional polynucleotide phosphatase/kinase; DNA 5'-kinase/3'-phosphatase; EIEE10; Homo sapiens polynucleotide kinase 3'-phosphatase (PNKP); MCSZ; phosphatase/kinase; PKNP; PNK; PNKP; Polynucleotide 3'-phosphatase; Polynucleotide 5'-hydroxyl-kinase; polynucleotide kinase 3'- phosphatase; polynucleotide kinase 3'-phosphatase; polynucleotide kinase-3'-phosphatase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11284 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q96T60 | |
| Pnkp | |
| Recombinant fragment corresponding to a region within amino acids 78 and 521 of Human PNK | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction