missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PNK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118170
This item is not returnable.
View return policy
Description
PNK Polyclonal Antibody for Western Blot, IP
Anti-human Polynucleotide Kinase Mouse Monoclonal Antibody recognizes polynucleotide kinase (PNK), a 57 kDa nuclear enzyme implicated in DNA strand repair processes that occur during DNA replication and recombination, or in response to DNA damaging agents. PNK targets the DNA 5'-hydroxy and 3'-phosphate termini for phosphorylation and dephosphorylation events. During strand breakage in human cells, normal PNK associates with other nuclear proteins to prepare the DNA termini for the enzymatic actions of DNA polymerase-beta and DNA ligase III. PNK is a key functional component in maintaining the genetic stability of human cells.
Specifications
| PNK | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q96T60 | |
| Pnkp | |
| E. coli-derived Human PNKP fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Pnkp | |
| 1810009G08Rik; 2'(3')-polynucleotidase; AOA4; bifunctional polynucleotide phosphatase/kinase; DNA 5'-kinase/3'-phosphatase; EIEE10; Homo sapiens polynucleotide kinase 3'-phosphatase (PNKP); MCSZ; phosphatase/kinase; PKNP; PNK; PNKP; Polynucleotide 3'-phosphatase; Polynucleotide 5'-hydroxyl-kinase; polynucleotide kinase 3'- phosphatase; polynucleotide kinase 3'-phosphatase; polynucleotide kinase-3'-phosphatase | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 11284 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction