missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12588-100ul
This item is not returnable.
View return policy
Description
PNK Polyclonal antibody specifically detects PNK in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| PNK | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-5uL/mg of uLysate | |
| DNA 5'-kinase/3'-phosphatase, EIEE10, Homo sapiens polynucleotide kinase 3'-phosphatase (PNKP), MCSZ, PNKbifunctional polynucleotide phosphatase/kinase, polynucleotide kinase 3'-phosphatase, Polynucleotide kinase-3'-phosphatase | |
| Produced in rabbits immunized with E. coli-derived Human PNK fragment | |
| 100 μg | |
| DNA Repair, Protein Phosphatase, Signal Transduction | |
| 11284 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction