missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies PneumaCult™-Ex Plus Medium
Description
- PneumaCult™-Ex Plus Medium is a defined, serum- and BPE-free cell culture medium that supports more expansion of primary human airway epithelial cells at each passage, compared to other commercially available expansion media
- This medium also supports at least two additional passages of cell expansion with better differentiation potential, defined as the ability to form a pseudostratified mucociliary epithelium at the air-liquid interface (ALI) using PneumaCult™-ALI Medium (Catalog #05001) or a cuboidal epithelium using PneumaCult™-ALI-S Medium (Catalog #05050)
- PneumaCult™-Ex Plus and either PneumaCult™-ALI or PneumaCult™-ALI-S constitute a fully integrated BPE-free culture system for in vitro human airway modeling
- This robust and defined system is a valuable tool for basic respiratory research, toxicity studies, and drug development
Specifications
Specifications
| Cell Type | Airway Cells |
| For Use With (Application) | Cell Culture; Expansion; Maintenance |
| Quantity | 1 Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?