missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies PneumaCult™-ALI Medium with 6.5 mm Transwell™ Inserts
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
- PneumaCult™-ALI Medium (Catalog #05001) is a serum- and BPE-free medium for the culture of human airway epithelial cells at the air-liquid interface (ALI)
- Airway epithelial cells cultured in PneumaCult™-ALI Medium undergo extensive mucociliary differentiation to form a pseudostratified epithelium that exhibits morphological and functional characteristics similar to those of the human airway in vivo
- PneumaCult™-ALI Medium is also available in a kit that includes 12 mm Transwell™ inserts (Catalog #05021) or 6.5 mm Transwell™ inserts (Catalog #05022)
- Together, PneumaCult™-ALI Medium and PneumaCult™-Ex Plus Medium (Catalog #05040) constitute a fully integrated BPE-free culture system for in vitro human airway modeling
- This robust and defined system is a valuable tool for basic respiratory research, toxicity studies, and drug development
Specifications
Specifications
| Cell Type | Airway Cells |
| For Use With (Application) | Cell Culture; Differentiation; Maintenance; Organoid Culture |
| Quantity | 1 Kit |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction