missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies PneumaCult™-ALI Medium with 12 mm Transwell™ Inserts

Product Code. 17138251
Encompass
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17138251

Brand: STEMCELL Technologies 05021

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Medium and Transwell™ inserts for human airway epithelial cells cultured at the air-liquid interface

  • PneumaCult™-ALI Medium (Catalog #05001) is a serum- and BPE-free medium for the culture of human airway epithelial cells at the air-liquid interface (ALI)
  • Airway epithelial cells cultured in PneumaCult™-ALI Medium undergo extensive mucociliary differentiation to form a pseudostratified epithelium that exhibits morphological and functional characteristics similar to those of the human airway in vivo
  • PneumaCult™-ALI Medium is also available in a kit that includes 12 mm Transwell™ inserts (Catalog #05021) or 6.5 mm Transwell™ inserts (Catalog #05022)
  • Together, PneumaCult™-ALI Medium and PneumaCult™-Ex Plus Medium (Catalog #05040) constitute a fully integrated BPE-free culture system for in vitro human airway modeling
  • This robust and defined system is a valuable tool for basic respiratory research, toxicity studies, and drug development

Specifications

Cell Type Airway Cells
For Use With (Application) Cell Culture; Differentiation; Maintenance; Organoid Culture
Quantity 1 Kit
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.