missing translation for 'onlineSavingsMsg'
Learn More

PND-1186, MedChemExpress

Product Code. 16488547
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488547 5 mg 5mg
16476356 100 mg 100mg
16425918 10 mg 10mg
16412067 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16488547 Supplier Medchem Express Supplier No. HY13917
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress PND-1186 (VS-4718) is a potent, highly-specific and reversible inhibitor of FAK with an IC50 of 1.5 nM. PND-1186 selectively promotes tumor cell apoptosis.

PND-1186 (VS-4718) is a potent, highly-specific and reversible inhibitor of FAK with an IC50 of 1.5 nM. PND-1186 selectively promotes tumor cell apoptosis.

Specifications

Chemical Name or Material PND-1186
CAS 1061353-68-1
Color White
Purity Grade Notes Research
Molecular Formula C25H26F3N5O3
Quantity 5 mg
Synonym VS-4718 SR-2516
Solubility Information DMSO : ≥ 34 mg/mL (67.80 mM)
SMILES O=C(NC)C1=CC=CC=C1NC2=CC(NC3=CC=C(N4CCOCC4)C=C3OC)=NC=C2C(F)(F)F
Molecular Weight (g/mol) 501.5
Formula Weight 501.5
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.