missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PND 1186

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
PND 1186 is a potent FAK inhibitor (IC50 values are 1.5 and 100 nM for recombinant FAK and FAK in breast carcinoma cells, respectively). Promotes caspase activation; triggering apoptosis. Inhibits 4T1 breast carcinoma subcutaneous tumor growth and breast carcinoma cell motility. Decreases spontaneous breast to lung metastasis in pre-clinical models. Orally bioavailable.
Specifications
Specifications
| Quantity | 10 mg |
| Molecular Formula | C25H26F3N5O3 |
| Formula Weight | 501.51 |
| Product Type | Inhibitor |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction