missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNCK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538710
This item is not returnable.
View return policy
Description
PNCK is a member of the calcium/calmodulin-dependent protein kinase family of protein serine/threonine kinases (see CAMK1; MIM 604998) (Gardner et al., 2000 [PubMed 10673339]).[supplied by OMIM, Mar 2008].
Specifications
| PNCK | |
| Polyclonal | |
| Unconjugated | |
| PNCK | |
| BSTK3, CaMK1b, caM kinase I beta, caM kinase IB, caM-KI beta, caMKI-beta, calcium/calmodulin-dependent protein kinase type 1B, pregcy up-regulated non-ubiquitously-expressed CaM kinase, pregcy upregulated non-ubiquitously expressed CaM kinase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 139728, 29660, 93843 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6P2M8, Q9QYK9, O70150 | |
| PNCK | |
| A synthetic peptide derived from the internal region of human CaMK1 beta | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction