missing translation for 'onlineSavingsMsg'
Learn More

PMVK/phosphomevalonate kinase Rabbit anti-Human, Clone: 14, Novus Biologicals™

Rabbit Monoclonal Antibody

£286.00 - £429.00

Specifications

Antigen PMVK/phosphomevalonate kinase
Clone 14
Dilution Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunoprecipitation 0.2-1 ul/mg of lysate
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18254867
View Documents
Novus Biologicals
NBP2-90297-50UL
50 μg
£286.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18287266
View Documents
Novus Biologicals
NBP2-90297-100UL
100 μg
£429.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PMVK/phosphomevalonate kinase Monoclonal specifically detects PMVK/phosphomevalonate kinase in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
Specifications

Specifications

PMVK/phosphomevalonate kinase
Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunoprecipitation 0.2-1 ul/mg of lysate
Monoclonal
Purified
RUO
0.2 um filtered solution in PBS
10654
IgG
Protein A purified
14
Western Blot, ELISA, Immunoprecipitation
Unconjugated
Rabbit
Human
hPMK, HUMPMKI, phosphomevalonate kinase, PMKA, PMKase, PMKEC 2.7.4.2, PMKI
Recombinant Human PMVK/phosphomevalonate kinase Protein
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.