missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PMS2 Rabbit anti-Human, Clone: PMS2/4373R, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-07615-100ug
This item is not returnable.
View return policy
Description
PMS2 Monoclonal specifically detects PMS2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| PMS2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin, Protein Array | |
| DNA mismatch repair protein PMS2, EC 3.1, H_DJ0042M02.9, HNPCC4postmeiotic segregation increased (S. cerevisiae) 2, mismatch repair endonuclease PMS2, PMS1 protein homolog 2, PMS2 postmeiotic segregation increased 2 (S. cerevisiae), PMSL2PMS2CL | |
| A synthetic peptide corresponding to residues within aa1-100 of human PMS2 (exact sequence is proprietary) | |
| 100 μg | |
| Cancer, Cell Cycle and Replication, DNA Repair, Mismatch Repair | |
| 5395 | |
| Store at 4°C. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| PMS2/4373R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction