missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PMS2 Polyclonal Antibody

Description
Purity is > 95% by SDS-PAGE.
PMS1 protein homologue 2 (PMS2) is a DNA mismatch repair protein. Defects in PMS2 are a cause of hereditary non-polyposis cancer (HNPCC). Human PMS2 is an amino acid protein of 862 amino acids.
Specifications
Specifications
| Antigen | PMS2 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | PMS2 |
| Gene Accession No. | P54278, P54279 |
| Gene Alias | AW555130; DNA mismatch repair protein PMS2; H_DJ0042M02.9; HNPCC4; mismatch repair endonuclease PMS2; MLH4; PMS1 homolog 2, mismatch repair protein; PMS1 homolog 2, mismatch repair system component; PMS1 protein homolog 2; PMS2; PMS2 postmeiotic segregation increased 2; PMS2 postmeiotic segregation increased 2 (S. cerevisiae); PMS2CL; PMSL2; postmeiotic segregation increased 2; postmeiotic segregation increased 2 (S. cerevisiae); postmeiotic segregation increased 2 nirs variant 6 |
| Gene Symbols | PMS2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?