missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PMS2 Antibody (PMS2/8374R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20297-20ug
This item is not returnable.
View return policy
Description
PMS2 Monoclonal antibody specifically detects PMS2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| PMS2 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| DNA mismatch repair protein PMS2, EC 3.1, H_DJ0042M02.9, HNPCC4postmeiotic segregation increased (S. cerevisiae) 2, mismatch repair endonuclease PMS2, PMS1 protein homolog 2, PMS2 postmeiotic segregation increased 2 (S. cerevisiae), PMSL2PMS2CL | |
| Recombinant fragment (around aa1-200) of human PMS2 protein (exact sequence is proprietary) | |
| 20 μg | |
| Cancer, Cell Cycle and Replication, DNA Repair, Mismatch Repair | |
| 5395 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| PMS2/8374R | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction