missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PMS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565087
This item is not returnable.
View return policy
Description
Immunogen sequence: ESVLIALENLM TTCYGPLPST NSYENNKTDV SAADIVLSKT AETDVLFNKV ESSGKNYSNV DTSVIPFQND MHNDESGKNT DDCLNHQISI GDFGYGHCSS Highest antigen sequence identity to the following orthologs - mouse 64%, rat 65%.
This gene encodes a protein belonging to the DNA mismatch repair mutL/hexB family. This protein is thought to be involved in the repair of DNA mismatches, and it can form heterodimers with MLH1, a known DNA mismatch repair protein. Mutations in this gene cause hereditary nonpolyposis colorectal cancer type 3 (HNPCC3) either alone or in combination with mutations in other genes involved in the HNPCC phenotype, which is also known as Lynch syndrome.
Specifications
| PMS1 | |
| Polyclonal | |
| Unconjugated | |
| PMS1 | |
| DNA mismatch repair protein PMS1; HNPCC3; hPMS1; human homolog of yeast mutL; mismatch repair gene PMSL1; MLH2; PMS1; PMS1 homolog 1, mismatch repair system component; PMS1 postmeiotic segregation increased 1; PMS1 protein homolog 1; PMSL1; postmeiotic segregation increased 1 (S. cerevisiae); rhabdomyosarcoma antigen MU-RMS-40.10B; rhabdomyosarcoma antigen MU-RMS-40.10E | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5378 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P54277 | |
| PMS1 | |
| Recombinant Human PMS1. Recombinant protein control fragment (Product #RP-105847). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction