missing translation for 'onlineSavingsMsg'
Learn More

PMS1 Antibody (PCRP-PMS1-2E11), Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18381123 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18381123 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18381123 Supplier Novus Biologicals Supplier No. NBP314142AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PMS1 Monoclonal antibody specifically detects PMS1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMS1
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone PCRP-PMS1-2E11
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DKFZp781M0253, FLJ98259, HNPCC3, hPMS1, human homolog of yeast mutL, mismatch repair gene PMSL1, PMS1 postmeiotic segregation increased 1 (S. cerevisiae), PMS1 protein homolog 1, PMSL1DNA mismatch repair protein PMS1, postmeiotic segregation increased (S. cerevisiae) 1, rhabdomyosarcoma antigen MU-RMS-40.10B, rhabdomyosarcoma antigen MU-RMS-40.10E
Host Species Mouse
Immunogen Recombinant full-length human PMS1 protein (Uniprot: P54277)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, Mismatch Repair, Ovarian Carcinoma Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5378
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.