missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PMPCB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16064-1-AP-20UL
This item is not returnable.
View return policy
Description
PMPCB is a member of the peptidase M16 family and encodes a protein with a zinc-binding motif. This protein is located in the mitochondrial matrix and catalyzes the cleavage of the leader peptides of precursor proteins newly imported into the mitochondria, though it only functions as part of a heterodimeric complex.Specifications
| PMPCB | |
| Polyclonal | |
| Unconjugated | |
| PMPCB | |
| Beta MPP, MPP11, MPPB, MPPP52, P 52, PMPCB | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64198, 73078, 9512 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunoprecipitation | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75439, Q03346, Q9CXT8 | |
| Pmpcb | |
| PMPCB Fusion Protein Ag8937 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction