Learn More
Invitrogen™ PMPCB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116547
Description
Positive Samples: LO2, HT-1080, Mouse kidney, Rat liver Immunogen sequence: GTKKRSQLDL ELEIENMGAH LNAYTSREQT VYYAKAFSKD LPRAVEILAD IIQNSTLGEA EIERERGVIL REMQEVETNL QEVVFDYLHA TAYQNTALGR TILGPTENIK SISRKDLVDY ITTHYKGPRI VLAAAGGVSH DELLDLAKFH FGDSLCTHKG EIPALPPCKF TGSEIRVRDD KMPLAHLAIA VEAVGWAHPD TICLMVANTL IGNWDRSFGG GMNLSSKLAQ LTCHGNLCHS FQSFNTSYTD TGLWGLYMVC ESSTVADMLH V.
PMPCB is a member of the peptidase M16 family and encodes a protein with a zinc-binding motif. This protein is located in the mitochondrial matrix and catalyzes the cleavage of the leader peptides of precursor proteins newly imported into the mitochondria, though it only functions as part of a heterodimeric complex.
Specifications
| PMPCB | |
| Polyclonal | |
| Unconjugated | |
| PMPCB | |
| 3110004O18Rik; Beta-MPP; mitochondrial processing peptidase beta subunit; mitochondrial-processing peptidase subunit beta; MPP11; MPPB; MPPP52; P-52; peptidase (mitochondrial processing) beta; peptidase, mitochondrial processing beta subunit; PMPCB | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 64198, 73078, 9512 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.46 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75439, Q03346, Q9CXT8 | |
| PMPCB | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 110-380 of human PMPCB (NP_0042702). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.