missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PMPA (NAALADase inhibitor)

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Highly potent and selective inhibitor of glutamate carboxypeptidase 2 (GCP II/N-acetylated α-linked dipeptidase/NAALADase) with a Ki value of 275 pM. Neuroprotective; protects against glutamate-mediated motor neuron degeneration and reduces volume of injury following middle cerebral artery occlusion (MCAO).
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 226.12 |
| Purity | >98% |
| Molecular Weight (g/mol) | 226.121 |
| Chemical Name or Material | PMPA (NAALADase inhibitor) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction