missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PMP70 Antibody (CL2524) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44465
This item is not returnable.
View return policy
Description
PMP70 Monoclonal antibody specifically detects PMP70 in Human samples. It is validated for Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence,Immunohistochemistry
Specifications
| PMP70 | |
| Monoclonal | |
| LYOPH | |
| Immunocytochemistry/ Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| 70 kDa peroxisomal membrane protein, ABC43, ATP-binding cassette, sub-family D (ALD), member 3, peroxisomal membrane protein 1 (70kD, Zellweger syndrome), Peroxisomal membrane protein-1 (70kD), PMP70ATP-binding cassette sub-family D member 3, PXMP1dJ824O18.1 (ATP-binding cassette, sub-family D (ALD), member 3 (PMP70, PXMP1)), ZWS2 | |
| This antibody was generated using a recombinant protein sequence of P28288, with the exact immunogen sequence remaining proprietary. | |
| 100 μL | |
| ABC Transporters, Cellular Markers, Lipid and Metabolism, Neuroscience, Peroxisome Markers, Signal Transduction | |
| 5825 | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence, Immunohistochemistry | |
| CL2524 | |
| Unconjugated | |
| Lyophilized in 5% Trehalose. | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL. | |
| Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction