missing translation for 'onlineSavingsMsg'
Learn More

PMP70 Antibody (2H6), Novus Biologicals™

Product Code. 18328588 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18328588 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18328588 Supplier Novus Biologicals Supplier No. H00005825M04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PMP70 Monoclonal antibody specifically detects PMP70 in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMP70
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2H6
Conjugate Unconjugated
Dilution ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_002849
Gene Alias 70 kDa peroxisomal membrane protein, ABC43, ATP-binding cassette, sub-family D (ALD), member 3, peroxisomal membrane protein 1 (70kD, Zellweger syndrome), Peroxisomal membrane protein-1 (70kD), PMP70ATP-binding cassette sub-family D member 3, PXMP1dJ824O18.1 (ATP-binding cassette, sub-family D (ALD), member 3 (PMP70, PXMP1)), ZWS2
Host Species Mouse
Immunogen ABCD3 (NP_002849.1, 351 a.a. ~ 449 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SELLEDYYQSGRMLLRMSQALGRIVLAGREMTRLAGFTARITELMQVLKDLNHGKYERTMVSQQEKGIEGVQVIPLIPGAGEIIIADNIIKFDHVPLAT
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline ABC Transporters, Cellular Markers, Lipid and Metabolism, Neuroscience, Peroxisome Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5825
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.