missing translation for 'onlineSavingsMsg'
Learn More

PMP22 Antibody (OTI5D1) [DyLight 488], Novus Biologicals™

Product Code. 30687626 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30687626 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30687626 Supplier Novus Biologicals Supplier No. NBP344912G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PMP22 Monoclonal antibody specifically detects PMP22 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMP22
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone OTI5D1
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias CMT1A, CMT1E, GAS3, GAS-3DSS, growth arrest-specific 3, Growth arrest-specific protein 3, HMSNIA, HNPP, MGC20769, peripheral myelin protein 22, PMP-22, Sp110
Host Species Mouse
Immunogen Full length human recombinant protein of human PMP22 (NP_000295) produced in HEK293T cell.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Glia Markers, Membrane Trafficking and Chaperones, Neuroscience, Oligodendrocyte Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5376
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.