missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Rack for Sedimentation Cones

Holds 2 Imhoff sedimentation cones made of plastic. Compact design and easy to carry, even with cones filled.

Brand:  BRAND™ 388060

117.84 GBP valid until 2024-04-26
Use promo code "21699" to get your promotional price.



Code : A0

Additional Details : Weight : 1.06000kg

 View more versions of this product

Product Code. 10606901

  • £136.22 / Each
In Stock
to see stock.
Add to basket

Description

Description

  • Holds 2 Imhoff sedimentation cones
  • Polymethyl methacrylate (PMMA)/Polypropylene (PP) construction
Specifications

Specifications

Imhoff Cone Rack
PMMA, Polypropylene
300 x 130 x 400mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers