missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PMM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556547
This item is not returnable.
View return policy
Description
Immunogen sequence: MAVTAQAARR KERVLCLFDV DGTLTPARQK IDPEVAAFLQ KLRSRVQIGV VGGSDY Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.
PMM1, also named as PMMH22, belongs to the eukaryotic PMM family. It catalyzes the second step in the conversion of fructose-6P to GDP-mannose is and is involved in the synthesis of the GDP-mannose and dolichol-phosphate-mannose required for a number of critical mannosyl transfer reactions. In addition, PMM1 may be responsible for the degradation of glucose-1, 6-bisphosphate in ischemic brain.
Specifications
| PMM1 | |
| Polyclonal | |
| Unconjugated | |
| Pmm1 | |
| brain glucose-1,6-bisphosphatase; C77612; Phosphomannomutase 1; PMM 1; Pmm1; PMMH22; PMMH-22; Sec53; Secp53 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29858, 300089, 5372 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O35621, Q92871 | |
| Pmm1 | |
| Recombinant protein corresponding to Human PMM1. Recombinant protein control fragment (Product #RP-95363). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction