missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PML Protein Rabbit anti-Human, Mouse, Rat, Clone: 2A0N10, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | PML Protein |
|---|---|
| Clone | 2A0N10 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18376756
|
Novus Biologicals
NBP3-15835-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18347852
|
Novus Biologicals
NBP3-15835-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PML Protein Monoclonal antibody specifically detects PML Protein in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| PML Protein | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Zinc Finger | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 5371 | |
| IgG | |
| Affinity purified |
| 2A0N10 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| MYLPP8675, promyelocytic leukemia, Promyelocytic leukemia protein, RING finger protein 71, RNF71probable transcription factor PML, TRIM19promyelocytic leukemia, inducer of, tripartite motif protein TRIM19, Tripartite motif-containing protein 19 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PML Protein (P29590). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title