missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PML Protein Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94515-0.02ml
This item is not returnable.
View return policy
Description
PML Protein Polyclonal antibody specifically detects PML Protein in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PML Protein | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| MYLPP8675, promyelocytic leukemia, Promyelocytic leukemia protein, RING finger protein 71, RNF71probable transcription factor PML, TRIM19promyelocytic leukemia, inducer of, tripartite motif protein TRIM19, Tripartite motif-containing protein 19 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 571-611 of human PML Protein (NP_150241.2). SSRELDDSSSESSDLQLEGPSTLRVLDENLADPQAEDRPLV | |
| 0.02 mL | |
| Cancer, Zinc Finger | |
| 5371 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction