missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PML Protein Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93086-0.1ml
This item is not returnable.
View return policy
Description
PML Protein Polyclonal antibody specifically detects PML Protein in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| PML Protein | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| MYLPP8675, promyelocytic leukemia, Promyelocytic leukemia protein, RING finger protein 71, RNF71probable transcription factor PML, TRIM19promyelocytic leukemia, inducer of, tripartite motif protein TRIM19, Tripartite motif-containing protein 19 | |
| A synthetic peptide corresponding to a sequence within amino acids 300-600 of human PML Protein (NP_150241.2). AVDARYQRDYEEMASRLGRLDAVLQRIRTGSALVQRMKCYASDQEVLDMHGFLRQALCRLRQEEPQSLQAAVRTDGFDEFKVRLQDLSSCITQGKDAAVSKKASPEAASTPRDPIDVDLPEEAERVKAQVQALGLAEAQPMAVVQSVPGAHPVPVYAFSIKGPSYGEDVSNTTTAQKRKCSQTQCPRKVIKMESEEGKEARLARSSPEQPRPSTSKAVSPPHLDGPPSPRSPVIGSEVFLPNSNHVASGAGEAEERVVVISSSEDSDAENSSSRELDDSSSESSDLQLEGPSTLRVLDENL | |
| 0.1 mL | |
| Cancer, Zinc Finger | |
| 5371 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction