missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PML Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580909
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human PML/RNF71/TRIM19.
The protein encoded by this gene is one of two human homologs of Saccharomyces cerevisiae Rad23, a protein involved in nucleotide excision repair (NER). This protein was shown to interact with, and elevate the nucleotide excision activity of 3-methyladenine-DNA glycosylase (MPG), which suggested a role in DNA damage recognition in base excision repair. This protein contains an N-terminal ubiquitin-like domain, which was reported to interact with 26S proteasome, as well as with ubiquitin protein ligase E6AP, and thus suggests that this protein may be involved in the ubiquitin mediated proteolytic pathway in cells.
Specifications
| PML | |
| Polyclonal | |
| Unconjugated | |
| PML | |
| 1200009E24Rik; AI661194; MYL; Pml; PML/RARA fusion; pml1; PP8675; probable transcription factor PML; promyelocytic leukemia; promyelocytic leukemia protein; promyelocytic leukemia, inducer of; protein PML; RGD1562602; RING finger protein 71; RNF71; TRIM19; tripartite motif protein TRIM19; tripartite motif-containing protein 19 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 5371 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with no preservative | |
| P29590 | |
| PML | |
| A synthetic peptide corresponding to the N-terminus of the Human PML/RNF71/TRIM19. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction