missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PMF1 Recombinant Rabbit Monoclonal Antibody (K01_2N96)
Description
PMF1 Recombinant Monoclonal Antibody for Western Blot
PMF1 is part of the MIS12 complex which is required for normal chromosome alignment and segregation and kinetochore formation during mitosis. It may act as a cotranscription partner of NFE2L2 involved in regulation of polyamine-induced transcription of SSAT.
Specifications
Specifications
| Antigen | PMF1 |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_2N96 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4 |
| Gene | PMF1 |
| Gene Accession No. | Q6P1K2, Q9CPV5 |
| Gene Alias | 2600009M07Rik; AL033286; AW060657; Est1p-like protein B (EST1B); Pmf1; PMF-1; polyamine modulated factor 1; polyamine-modulated factor 1; RP11-54H19.4; Unknown (protein for MGC:134238) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?