missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PMEPA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16521-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a transmembrane protein that contains a Smad interacting motif (SIM). Expression of this gene is induced by androgens and transforming growth factor beta, and the encoded protein suppresses the androgen receptor and transforming growth factor beta signaling pathways though interactions with Smad proteins. Overexpression of this gene may play a role in multiple types of cancer. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Specifications
| PMEPA1 | |
| Polyclonal | |
| Unconjugated | |
| PMEPA1 | |
| PMEPA1, STAG1, TMEPAI | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 311676, 56937, 65112 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q969W9, Q9D7R2 | |
| Pmepa1 | |
| PMEPA1 Fusion Protein Ag9744 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction