missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PMEPA1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16607714
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16607714 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16607714 Supplier Invitrogen™ Supplier No. PA5101231

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PMEPA1.

This gene encodes a transmembrane protein that contains a Smad interacting motif (SIM). Expression of this gene is induced by androgens and transforming growth factor beta, and the encoded protein suppresses the androgen receptor and transforming growth factor beta signaling pathways though interactions with Smad proteins. Overexpression of this gene may play a role in multiple types of cancer. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMEPA1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PMEPA1
Gene Accession No. Q969W9, Q9D7R2
Gene Alias 2210418I02Rik; AW455466; Erg1.2; N4wbp4; Nedd4 WW binding protein 4; NEDD4 WW domain-binding protein 4; PMEPA1; Prostate transmembrane protein androgen induced 1; prostate transmembrane protein, androgen induced 1; protein TMEPAI; Solid tumor-associated 1 protein; STAG1; Tmepai; transmembrane prostate androgen-induced protein; transmembrane, prostate androgen induced RNA
Gene Symbols PMEPA1
Host Species Rabbit
Immunogen A synthesized peptide derived from human PMEPA1(Accession Q969W9), corresponding to amino acid residues F72-P122.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56937, 65112
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.