missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PMEL17/SILV Antibody (NKI-beteb), Novus Biologicals™
Mouse Monoclonal Antibody
£226.00 - £469.00
Specifications
| Antigen | PMEL17/SILV |
|---|---|
| Clone | NKI-beteb |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array, Flow (Intracellular) |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array, Flow Cytometry, Immunohistochemistry (Frozen) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18492122
|
Novus Biologicals
NBP2-29407-20ug |
20 ug |
£226.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18729193
|
Novus Biologicals
NBP2-29407 |
£469.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
PMEL17/SILV Monoclonal specifically detects PMEL17/SILV in Human, Equine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array, Flow (Intracellular).Specifications
| PMEL17/SILV | |
| 0.2 mg/ml | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array, Flow Cytometry, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Human, Equine | |
| D12S53EP1, gp100, ME20, ME20-M, melanocyte protein mel 17, Melanocyte protein Pmel 17, Melanocytes lineage-specific antigen GP100, Melanoma-associated ME20 antigen, melanosomal matrix protein17, PMEL17P100, premelanosome proteinME20M, SI, SIL, silver (mouse homolog) like, silver homolog (mouse), Silver locus protein homolog, silver, mouse, homolog of, SILVPmel17 | |
| PMEL | |
| IgG2b κ | |
| Protein A or G purified | |
| By immunohistochemistry, it specifically recognizes a protein in melanocytes and melanomas. This monoclonal antibody reacts with junctional and blue nevus cells and variably with fetal and neonatal melanocytes. Intradermal nevi, normal adult melanocytes, and non-melanocytic cells are negative. It does not stain tumor cells of epithelial, lymphoid, glial, or mesenchymal origin. This Mab labels formalin-fixed, paraffin-embedded melanomas and other tumors showing melanocytic differentiation. |
| NKI-beteb | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array, Flow (Intracellular) | |
| Monoclonal | |
| Purified | |
| RUO | |
| P40967 | |
| 6490 | |
| Lymph node pigmented melanoma metastases extract was used as the immunogen for the gp100 NKI-beteb antibody | |
| Primary | |
| Store at 4C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title