missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PMEL17/SILV Antibody (Genentech anti-PMEL17) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
£162.00 - £695.00
Specifications
| Antigen | PMEL17/SILV |
|---|---|
| Clone | Genentech anti-PMEL17 |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30044097
|
Novus Biologicals™
NBP3-28129-1mg |
1 mg |
£695.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30048732
|
Novus Biologicals™
NBP3-28129-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30047020
|
Novus Biologicals™
NBP3-28129-100ug |
100 μg |
£257.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PMEL17/SILV Monoclonal antibody specifically detects PMEL17/SILV in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| PMEL17/SILV | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Human | |
| P40967 | |
| 6490 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
| Genentech anti-PMEL17 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| D12S53EP1, gp100, ME20, ME20-M, melanocyte protein mel 17, Melanocyte protein Pmel 17, Melanocytes lineage-specific antigen GP100, Melanoma-associated ME20 antigen, melanosomal matrix protein17, PMEL17P100, premelanosome proteinME20M, SI, SIL, silver (mouse homolog) like, silver homolog (mouse), Silver locus protein homolog, silver, mouse, homolog of, SILVPmel17 | |
| PMEL | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title