missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PMCA4 Polyclonal antibody specifically detects PMCA4 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | PMCA4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS (pH 7.4) |
| Gene Accession No. | NP_001001396.1 |
| Gene Alias | ATP2B2plasma membrane calcium ATPase, ATPase, Ca++ transporting, plasma membrane 4, DKFZp686G08106, DKFZp686M088, EC 3.6.3, EC 3.6.3.8, Matrix-remodeling-associated protein 1, matrix-remodelling associated 1, MXRA1, Plasma membrane calcium ATPase isoform 4, plasma membrane calcium pump, Plasma membrane calcium pump isoform 4, plasma membrane calcium-transporting ATPase 4, PMCA4b, PMCA4PMCA4x, sarcolemmal calcium pump |
| Host Species | Rabbit |
| Immunogen | ATP2B4 (NP_001001396.1, 1 a.a. - 1170 a.a.) full-length human protein. MTNPSDRVLPANSMAESREGDFGCTVMELRKLMELRSRDALTQINVHYGGVQNLCSRLKTSPVEGLSGNPADLEKRRQVFGHNVIPPKKPKTFLELVWEALQDVTLIILEIAAIISLVLSFYRPAGEENELCGQVATTPEDENEAQAGWIEGAAILFSVIIVVLVTAFNDWSKEKQFRGLQCRIEQEQKFSIIRNGQLIQLPVAEIVVGDIAQVKYGDLLPADGILIQGNDLKIDESSLTGESDHVKKSLDKDPMLLSGTHVMEGSGRMVVTAVGVNSQTGIILTLLGVNEDDEGEKKKKGKKQGVPENRNKAKTQDGVALEIQPLNSQEGIDNEEKDKKAVKVPKKEKSVLQGKLTRLAVQIGKAGLLMSALTVFILILYFVIDNFVINRRPWLPECTPIYIQYFVKFFIIGITVLVVAVPEGLPLAVTISLAYSVKKMMKDNNLVRHLDACETMGNATAICSDKTGTLTMNRMTVVQAYIGGIHYRQIPSPDVFLPKVLDLIVNGISINSAYTSKILPPEKEGGLPRQVGNKTECALLGFVTDLKQDYQAVRNEVPEEKLYKVYTFNSVRKSMSTVIRNPNGGFRMYSKGASEIILRKCNRILDRKGEAVPFKNKDRDDMVRTVIEPMACDGLRTICIAYRDFDDTEPSWDNENEILTELTCIAVVGIEDPVRPEVPDAIAKCKQAGITVRMVTGDNINTARAIATKCGILTPGDDFLCLEGKEFNRLIRNEKGEVEQEKLDKIWPKLRVLARSSPTDKHTLVKGIIDSTVGEHRQVVAVTGDGTNDGPALKKADVGFAMGIAGTDVAKEASDIILTDDNFTSIVKAVMWGRNVYDSISKFLQFQLTVNVVAVIVAFTGACITQDSPLKAVQMLWVNLIMDTFASLALATEPPTESLLKRRPYGRNKPLISRTMMKNILGHAFYQLIVIFILVFAGEKFFDIDSGRKAPLHSPPSQHYTIVFNTFVLMQLFNEINSRKIHGEKNVFSGIYRNIIFCSVVLGTFICQIFIVEFGGKPFSCTSLSLSQWLWCLFIGIGELLWGQFISAIPTRSLKFLKEAGHGTTKEEITKDAEGLDEIDHAEMELRRGQILWFRGLNRIQTQIDVINTFQTGASFKGVLRRQNMGQHLDVKLVPSSSYVAVAPVKSSPTTSVPAVSSPPMGNQSGQSVP |
| Purification Method | Protein G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?