missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PMCA1 ATPase Recombinant Rabbit Monoclonal Antibody (1X7X6)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558104
This item is not returnable.
View return policy
Description
Immunogen sequence: MGDMANNSVA YSGVKNSLKE ANHDGDFGIT LAELRALMEL RSTDALRKIQ ESYGDVYGIC TKLKTSPNEG LSGNPADLER REAVFGKNFI PPKKPKTFLQ.
The calcium pump of the plasma membrane, termed PMCA ATPase, pumps calcium from the cytosol to the extracellular space. This membrane-bound enzyme is related to a number of other ATPases including the SERCA ATPase and the sodium/potassium pump. There are four different genes encoding PMCA ATPase and studies have revealed 20 isoforms of the pump generated by alternate splicing of the primary gene products. mRNA distribution studies show that gene products 1 and 4 are transcribed in most tissues, however, products 2 and 3 are more tissue specific. Transcription of the splicing variants has also been found to be tissue specific. In the pancreas, where insulin secretion is calcium dependent, the beta cells only express the 4b isoform, however the alpha and gamma cells express both 4a and 4b isoforms. Studies have also shown that different splice variants have different affinities for calcium and calmodulin.
Specifications
| PMCA1 ATPase | |
| Recombinant Monoclonal | |
| 1.74 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| G5E829, P11505, P20020 | |
| ATP2B1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ATP2B1 (NP_001673.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 1X7X6 | |
| Unconjugated | |
| ATP2B1 | |
| 2810442I22Rik; Atp2b1; ATPase plasma membrane Ca2+ transporting 1; ATPase, Ca++ transporting, plasma membrane 1; E130111D10Rik; plasma membrane Ca2+ pump (PMCA1b); plasma membrane calcium ATPase; Plasma membrane calcium ATPase isoform 1; plasma membrane calcium pump; Plasma membrane calcium pump isoform 1; Plasma membrane calcium-transporting ATPase 1; PMCA1; Pmca1a; Pmca1b; Pmca1c; PMCA1kb | |
| Rabbit | |
| Protein A | |
| RUO | |
| 29598, 490, 67972 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction