missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PM20D1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546590
This item is not returnable.
View return policy
Description
Peptide sequence: SMGPRSGEHQ RASRIPSQFS KEERVAMKEA LKGAIQIPTV TFSSEKSNTT Sequence homology: Guinea Pig: 91%; Human: 100%.
Bidirectional N-fatty-acyl amino acid synthase/hydrolase that regulates the production of N-fatty-acyl amino acids. These metabolites are endogenous chemical uncouplers of mitochondrial respiration. In an UCP1-independent manner, maybe through interaction with mitochondrial transporters, they promote proton leakage into the mitochondrial matrix. Thereby, this secreted protein may indirectly regulate the bodily dissipation of chemical energy as heat through thermogenic respiration. [UniProt].
Specifications
| PM20D1 | |
| Polyclonal | |
| Unconjugated | |
| Pm20d1 | |
| Cps1; N-fatty-acyl-amino acid synthase/hydrolase PM20D1; peptidase M20 domain containing 1; peptidase M20 domain-containing protein 1; PM20D1; probable carboxypeptidase PM20D1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 148811 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6GTS8 | |
| Pm20d1 | |
| Synthetic peptide directed towards the N-terminal of human PM20D1 (aa 25-74). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction