missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLZF Monoclonal Antibody (Mags.21F7), Brilliant Violet™ 711, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£217.00 - £443.00
Specifications
| Antigen | PLZF |
|---|---|
| Clone | Mags.21F7 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17914681
|
Invitrogen™
407-9320-80 |
25 μg |
£217.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17964711
|
Invitrogen™
407-9320-82 |
100 μg |
£443.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene is a member of the Krueppel C2H2-type zinc-finger protein family and encodes a zinc finger transcription factor that contains nine Kruppel-type zinc finger domains at the carboxyl terminus. This protein is located in the nucleus, is involved in cell cycle progression, and interacts with a histone deacetylase. Specific instances of aberrant gene rearrangement at this locus have been associated with acute promyelocytic leukemia (APL). Alternate transcriptional splice variants have been characterized.Specifications
| PLZF | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| AI467657; Green's luxoid; lu; luxoid; Lx; PLZF; Polydactyly-luxate syndrome; promyelocytic leukaemia zinc finger; promyelocytic leukemia zinc finger; Promyelocytic leukemia zinc finger protein; ZBTB16; Zfp145; zinc finger and BTB domain containing 16; zinc finger and BTB domain containing 16 protein; zinc finger and BTB domain-containing protein 16; Zinc finger protein 145; zinc finger protein 145 (Kruppel-like, expressed in promyelocytic leukemia); zinc finger protein PLZF; ZNF145 | |
| ZBTB16 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| Mags.21F7 | |
| Flow Cytometry | |
| Brilliant Violet 711 | |
| Mouse | |
| Human, Mouse | |
| Q05516 | |
| 235320, 7704 | |
| IgG1 κ | |
| Affinity chromatography | |
| ZBTB16 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title