missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLZF Monoclonal Antibody (816416)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561945
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene is a member of the Krueppel C2H2-type zinc-finger protein family and encodes a zinc finger transcription factor that contains nine Kruppel-type zinc finger domains at the carboxyl terminus. This protein is located in the nucleus, is involved in cell cycle progression, and interacts with a histone deacetylase. Specific instances of aberrant gene rearrangement at this locus have been associated with acute promyelocytic leukemia (APL). Alternate transcriptional splice variants have been characterized.
Specifications
| PLZF | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| AI467657; Green's luxoid; lu; luxoid; Lx; PLZF; Polydactyly-luxate syndrome; promyelocytic leukaemia zinc finger; promyelocytic leukemia zinc finger; Promyelocytic leukemia zinc finger protein; ZBTB16; Zfp145; zinc finger and BTB domain containing 16; zinc finger and BTB domain containing 16 protein; zinc finger and BTB domain-containing protein 16; Zinc finger protein 145; zinc finger protein 145 (Kruppel-like, expressed in promyelocytic leukemia); zinc finger protein PLZF; ZNF145 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 235320 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 816416 | |
| Unconjugated | |
| ZBTB16 | |
| ZBTB16 | |
| E. coli-derived recombinant mouse PLZF, Met1-Gln254. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction