Learn More
Invitrogen™ PLZF Monoclonal Antibody (5B3)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515667
Description
MA5-15667 targets ZBTB16 in indirect ELISA, IF and WB applications and shows reactivity with Human samples. The MA5-15667 immunogen is purified recombinant fragment of human ZBTB16 expressed in E. Coli.. MA5-15667 detects ZBTB16 which has a predicted molecular weight of approximately 74kDa.
This gene is a member of the Krueppel C2H2-type zinc-finger protein family and encodes a zinc finger transcription factor that contains nine Kruppel-type zinc finger domains at the carboxyl terminus. This protein is located in the nucleus, is involved in cell cycle progression, and interacts with a histone deacetylase. Specific instances of aberrant gene rearrangement at this locus have been associated with acute promyelocytic leukemia (APL). Alternate transcriptional splice variants have been characterized.
Specifications
| PLZF | |
| Monoclonal | |
| Conc. Not Determined | |
| Ascites with 0.03% sodium azide | |
| Q05516 | |
| ZBTB16 | |
| Purified recombinant fragment of human ZBTB16 expressed in E. Coli. | |
| RUO | |
| 7704 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 5B3 | |
| Unconjugated | |
| ZBTB16 | |
| AI467657; Green's luxoid; lu; luxoid; Lx; PLZF; Polydactyly-luxate syndrome; promyelocytic leukaemia zinc finger; promyelocytic leukemia zinc finger; Promyelocytic leukemia zinc finger protein; ZBTB16; Zfp145; zinc finger and BTB domain containing 16; zinc finger and BTB domain containing 16 protein; zinc finger and BTB domain-containing protein 16; Zinc finger protein 145; zinc finger protein 145 (Kruppel-like, expressed in promyelocytic leukemia); zinc finger protein PLZF; ZNF145 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.