missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLXND1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111972
This item is not returnable.
View return policy
Description
PLXND1 Polyclonal Antibody for Western Blot, IHC (F)
Cell surface receptor for SEMA4A and for class 3 semaphorins, such as SEMA3A, SEMA3C and SEMA3E. Plays an important role in cell-cell signaling, and in regulating the migration of a wide spectrum of cell types. Regulates the migration of thymocytes in the medulla. Regulates endothelial cell migration. Plays an important role in ensuring the specificity of synapse formation. Required for normal development of the heart and vasculature. Mediates anti-angiogenic signaling in response to SEMA3E. [UniProt].
Specifications
| PLXND1 | |
| Polyclonal | |
| Unconjugated | |
| PLXND1 | |
| 6230425C21Rik; b2b1863Clo; b2b553Clo; KIAA0620; MGC75353; PLEXD1; plexin D1; plexin-D1; Plxnd1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23129, 67784 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen), Western Blot | |
| 0.95 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q3UH93, Q9Y4D7 | |
| PLXND1 | |
| Synthetic peptide encompassing a sequence within the Extracellular domain of human Plexin D1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction