missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLXND1 Monoclonal Antibody (752815)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561395
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
Cell surface receptor for SEMA4A and for class 3 semaphorins, such as SEMA3A, SEMA3C and SEMA3E. Plays an important role in cell-cell signaling, and in regulating the migration of a wide spectrum of cell types. Regulates the migration of thymocytes in the medulla. Regulates endothelial cell migration. Plays an important role in ensuring the specificity of synapse formation. Required for normal development of the heart and vasculature. Mediates anti-angiogenic signaling in response to SEMA3E. [UniProt].
Specifications
| PLXND1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9Y4D7 | |
| PLXND1 | |
| Mouse myeloma cell line NS0-derived recombinant human Plexin D1, Leu47-Ala1271. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 752815 | |
| Unconjugated | |
| PLXND1 | |
| 6230425C21Rik; b2b1863Clo; b2b553Clo; KIAA0620; MGC75353; PLEXD1; plexin D1; plexin-D1; Plxnd1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 23129 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction