missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PLXNC1 Monoclonal Antibody (2H11C11)
GREENER_CHOICE

Product Code. 17995591
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17995591 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17995591 Supplier Invitrogen™ Supplier No. MA548682

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PLXNC1 Monoclonal Antibody for Western Blot, IHC, Flow, ELISA

Receptor for SEMA7A, for smallpox semaphorin A39R, vaccinia virus semaphorin A39R and for herpesvirus Sema protein. Binding of semaphorins triggers cellular responses leading to the rearrangement of the cytoskeleton and to secretion of IL6 and IL8. {ECO:0000250, ECO:0000269; PubMed:20727575}.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLXNC1
Applications ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2H11C11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene PLXNC1
Gene Accession No. O60486
Gene Alias 2510048K12Rik; AW742158; CD232; CD232 antigen; plexin (semaphorin receptor); plexin C1; plexin-C1; PLXNC1; PLXN-C1; receptor for viral semaphorin protein; receptor for virally-encoded semaphorin; Vespr; VESPRplexin-C1; viral-encoded semaphorin protein receptor; virus-encoded semaphorin protein receptor
Gene Symbols PLXNC1
Host Species Mouse
Immunogen Purified recombinant fragment of human CD232 (AA: extra 35-234) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10154
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.