missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLXNB1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23795-1-AP-150UL
This item is not returnable.
View return policy
Description
Plexin B1 is a protein of the plexin family that in humans is encoded by the PLXNB1gene. Receptor for SEMA4D. Plays a role in RHOA activation and subsequent changes of the actin cytoskeleton. Plays a role in axon guidance, invasive growth and cell migration.Specifications
| PLXNB1 | |
| Polyclonal | |
| Unconjugated | |
| PLXNB1 | |
| KIAA0407, plexin B1, PLXN5, PLXNB1, Semaphorin receptor SEP, SEP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 235611, 5364 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43157, Q8CJH3 | |
| PLXNB1 | |
| PLXNB1 Fusion Protein Ag20719 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction