missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLXNA4 Monoclonal Antibody (F.566.2)
Rabbit Monoclonal Antibody
Brand: Invitrogen™ MA514795
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
PLXNA4 is a protein coding gene. Coreceptor for SEMA3A. Necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. Plays a role in axon guidance in the developing nervous system. Class 3 semaphorins bind to a complex composed of a neuropilin and a plexin. The plexin modulates the affinity of the complex for specific semaphorins, and its cytoplasmic domain is required for the activation of down-stream signaling events in the cytoplasm.
Specifications
| PLXNA4 | |
| Monoclonal | |
| 155 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
| Q80UG2, Q9HCM2 | |
| Plxna4 | |
| GST-fusion protein corresponding to residues from the human sequence of Plexin A4. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| F.566.2 | |
| Unconjugated | |
| Plxna4 | |
| 9330117B14; FAYV2820; Kiaa1550; mKIAA1550; OTTHUMP00000207245; PLEXA4; plexin A4; plexin A4, A; plexin-A4; Plxa4; Plxna4; Plxna4a; PLXNA4B; PRO34003; UNQ2820/PRO34003 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 243743, 312213, 91584 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction