missing translation for 'onlineSavingsMsg'
Learn More

PLX7904, MedChemExpress

Product Code. 16487576
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16487576

Brand: Medchem Express HY18997

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PLX7904 is a potent and selective BRAF inhibitor, with IC50 of appr 5 nM against BRAFV600E in mutant RAS expressing cells.

PLX7904 is a potent and selective BRAF inhibitor, with IC50 of appr 5 nM against BRAFV600E in mutant RAS expressing cells.

Specifications

Chemical Name or Material PLX7904
CAS 1393465-84-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H22F2N6O3S
Quantity 100 mg
Synonym PB04
Solubility Information DMSO : ≥ 30 mg/mL (58.53 mM)
SMILES O=S(NC1=CC=C(F)C(C(C2=CNC3=NC=C(C4=CN=C(C5CC5)N=C4)C=C32)=O)=C1F)(N(CC)C)=O
Molecular Weight (g/mol) 512.53
Formula Weight 512.53
Percent Purity 98.34%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.